Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117533.2 | XP_013973008.1 | 3 | 309 | 133 | |
| XM_022424732.1 | XP_022280440.1 | 3 | 317 | 226 | ||
| XM_022424733.1 | XP_022280441.1 | 3 | 318 | 215 | ||
| XM_022424734.1 | XP_022280442.1 | 7 | 988 | 226 | ||
| XM_022424735.1 | XP_022280443.1 | 7 | 888 | 226 | ||
| XM_022424736.1 | XP_022280444.1 | 7 | 720 | 226 | ||
| XM_022424737.1 | XP_022280445.1 | 8 | 1186 | 226 | ||
| XM_022424738.1 | XP_022280446.1 | 6 | 849 | 226 | ||
| XM_022424739.1 | XP_022280447.1 | 6 | 535 | 226 | ||
| XM_022424740.1 | XP_022280448.1 | 3 | 172 | 226 | ||
| XM_022424741.1 | XP_022280449.1 | 5 | 273 | 226 | ||
| XM_022424742.1 | XP_022280450.1 | 4 | 665 | 226 | ||
| XM_022424743.1 | XP_022280451.1 | 3 | 324 | 106 | ||
| XM_022424744.1 | XP_022280452.1 | 3 | 350 | 226 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ACYP2
Genomic Map