Assay Details
| Gene Symbol: NACC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625132.3 | XP_005625189.1 | 6 | 1129 | 96 | |
| XM_014116944.2 | XP_013972419.1 | 6 | 1144 | 96 | ||
| XM_022423604.1 | XP_022279312.1 | 6 | 1881 | 96 | ||
| XM_022423605.1 | XP_022279313.1 | 6 | 1147 | 96 | ||
| XM_548376.7 | XP_548376.2 | 6 | 1619 | 96 | ||
More Information
Set Membership: |
|

NACC2
Genomic Map