Assay Details
| Gene Symbol: RBM46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629370.3 | XP_005629427.1 | 3 | 307 | 148 | |
| XM_022403533.1 | XP_022259241.1 | 3 | 345 | 148 | ||
| XM_022403534.1 | XP_022259242.1 | 3 | 345 | 148 | ||
| XM_022403535.1 | XP_022259243.1 | 3 | 345 | 148 | ||
| XM_022403536.1 | XP_022259244.1 | 3 | 345 | 148 | ||
| XM_532699.5 | XP_532699.1 | 3 | 345 | 148 | ||
More Information
Set Membership: |
|

RBM46
Genomic Map