Assay Details
Gene Symbol: ZNRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014118908.2 | XP_013974383.1 | 4 | 574 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
