Assay Details
| Gene Symbol: LOC477540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_534735.5 | XP_534735.1 | 1 - 2 | 154 | 158 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC477540| Gene Symbol: LOC477540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_534735.5 | XP_534735.1 | 1 - 2 | 154 | 158 | |
Set Membership: |
|