Assay Details
| Gene Symbol: NR2C2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_847056.5 | XP_852149.1 | 2 | 344 | 191 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR2C2AP| Gene Symbol: NR2C2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_847056.5 | XP_852149.1 | 2 | 344 | 191 | |
Set Membership: |
|