Assay Details
| Gene Symbol: GPR142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_540406.4 | XP_540406.1 | 1350 | 108 | ||
| GenBank mRNA | KT749969.1 | - | 903 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GPR142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_540406.4 | XP_540406.1 | 1350 | 108 | ||
| GenBank mRNA | KT749969.1 | - | 903 | 108 | ||
Set Membership: |
|