Assay Details
| Gene Symbol: PKIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433272.4 | XP_003433320.1 | 3 | 322 | 125 | |
| XM_005635092.3 | XP_005635149.1 | 4 | 496 | 125 | ||
| XM_005635093.3 | XP_005635150.1 | 4 | 386 | 125 | ||
| XM_005635096.3 | XP_005635153.1 | 4 | 548 | 125 | ||
| XM_022409467.1 | XP_022265175.1 | 4 | 486 | 125 | ||
| XM_848508.3 | XP_853601.1 | 3 | 508 | 125 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PKIG
Genomic Map