Assay Details
Gene Symbol: TAFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022406461.1 | XP_022262169.1 | 495 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
