Assay Details
| Gene Symbol: HNRNPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640349.3 | XP_005640406.1 | 1 - 2 | 158 | 181 | |
| XM_005640350.3 | XP_005640407.1 | 1 - 2 | 158 | 181 | ||
| XM_543761.6 | XP_543761.2 | 1 - 2 | 158 | 181 | ||
More Information
Set Membership: |
|

HNRNPA3
Genomic Map