Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640820.3 | XP_005640877.1 | 2 | 710 | 98 | |
| XM_005640821.3 | XP_005640878.1 | 2 | 867 | 98 | ||
| XM_014111269.2 | XP_013966744.1 | 3 | 785 | 98 | ||
| XM_536117.6 | XP_536117.1 | 2 | 553 | 98 | ||
More Information
Set Membership: |
|

BRINP3
Genomic Map