Assay Details
| Gene Symbol: LOC474655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626440.3 | XP_005626497.1 | 2 | 692 | 130 | |
| XM_005626442.3 | XP_005626499.1 | 2 | 696 | 130 | ||
| XM_022425197.1 | XP_022280905.1 | 3 | 804 | 130 | ||
| XM_022425199.1 | XP_022280907.1 | 3 | 799 | 130 | ||
| XM_022425200.1 | XP_022280908.1 | 2 | 698 | 130 | ||
| XM_022425201.1 | XP_022280909.1 | 2 | 698 | 130 | ||
| XM_022425202.1 | XP_022280910.1 | 2 | 697 | 130 | ||
More Information
Set Membership: |
|

LOC474655
Genomic Map