Assay Details
| Gene Symbol: SKIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639910.3 | XP_005639967.1 | 2 | 1562 | 160 | |
| XM_005639911.3 | XP_005639968.1 | 2 | 1268 | 160 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SKIL
Genomic Map