Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115788.1 | XP_013971263.1 | 3 - 4 | 522 | 141 | |
| XM_014115790.1 | XP_013971265.1 | 3 - 4 | 522 | 141 | ||
| XM_014115791.1 | XP_013971266.1 | 3 - 4 | 522 | 141 | ||
| XM_014115792.1 | XP_013971267.1 | 3 - 4 | 501 | 141 | ||
| XM_537387.5 | XP_537387.3 | 3 - 4 | 501 | 141 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CHMP4A
Genomic Map