Assay Details
| Gene Symbol: RNF19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627829.3 | XP_005627886.1 | 1738 | 77 | ||
| XM_022426584.1 | XP_022282292.1 | 1465 | 77 | |||
More Information
Set Membership: |
|

RNF19A
Genomic Map