Assay Details
| Gene Symbol: RRM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627840.2 | XP_005627897.1 | 410 | 141 | ||
| XM_022426599.1 | XP_022282307.1 | 504 | 141 | |||
| XM_022426600.1 | XP_022282308.1 | 504 | 141 | |||
| XM_539109.6 | XP_539109.4 | 503 | 141 | |||
More Information
Set Membership: |
|

RRM2B
Genomic Map