Assay Details
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620830.3 | XP_005620887.1 | 4 - 5 | 543 | 165 | |
XM_014114104.2 | XP_013969579.1 | 4 - 5 | 543 | 165 | ||
XM_014114105.2 | XP_013969580.1 | 4 - 5 | 543 | 165 | ||
XM_014114106.2 | XP_013969581.1 | 2 - 3 | 273 | 165 | ||
XM_022419339.1 | XP_022275047.1 | 4 - 5 | 543 | 165 | ||
XM_022419340.1 | XP_022275048.1 | 4 - 5 | 543 | 165 | ||
XM_022419342.1 | XP_022275050.1 | 4 - 5 | 543 | 165 | ||
XM_022419343.1 | XP_022275051.1 | 2 - 3 | 273 | 165 | ||
XM_022419344.1 | XP_022275052.1 | 4 - 5 | 543 | 165 | ||
XM_022419345.1 | XP_022275053.1 | 4 - 5 | 543 | 165 | ||
XM_536819.6 | XP_536819.3 | 4 - 5 | 543 | 165 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
