Assay Details
| Gene Symbol: ZNF148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639586.3 | XP_005639643.1 | 6 - 7 | 1181 | 94 | |
| XM_022414014.1 | XP_022269722.1 | 5 - 6 | 783 | 94 | ||
More Information
Set Membership: |
|

ZNF148
Genomic Map