Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629853.2 | XP_005629910.1 | 2379 | 89 | ||
XM_005629854.2 | XP_005629911.1 | 2379 | 89 | |||
XM_005629855.2 | XP_005629912.1 | 2379 | 89 | |||
XM_005629856.2 | XP_005629913.1 | 2379 | 89 | |||
XM_014120026.2 | XP_013975501.1 | 2379 | 89 |
