Assay Details
| Gene Symbol: EGLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616548.3 | XP_005616605.1 | 4 - 5 | 1481 | 135 | |
| XM_005616549.3 | XP_005616606.1 | 4 - 5 | 1550 | 135 | ||
| XM_005616550.3 | XP_005616607.1 | 4 - 5 | 1452 | 135 | ||
| XM_541609.6 | XP_541609.1 | 4 - 5 | 1432 | 135 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EGLN2
Genomic Map