Assay Details
| Gene Symbol: CDKN2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122396.2 | XP_013977871.1 | 1 - 2 | 306 | 131 | |
| XM_848853.5 | XP_853946.1 | 2 - 3 | 277 | 131 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDKN2D
Genomic Map