Assay Details
Gene Symbol: COLEC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623024.3 | XP_005623081.1 | 7 - 8 | 2083 | 160 | |
XM_022421537.1 | XP_022277245.1 | 7 - 8 | 2101 | 160 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
