Assay Details
| Gene Symbol: ATP6V1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014376.1 | NP_001014398.1 | 2 - 3 | 184 | 158 | |
| XM_005627044.1 | XP_005627101.1 | 2 - 3 | 298 | 158 | ||
| XM_022425721.1 | XP_022281429.1 | 2 - 3 | 412 | 158 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_005627044.1, XM_022425721.1 transcripts.
This assay is in the 5'UTR region of the XM_005627044.1, XM_022425721.1 transcripts.
Set Membership: |
|

ATP6V1G2
Genomic Map