Assay Details
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639320.3 | XP_005639377.1 | 2 - 3 | 440 | 150 | |
XM_022413651.1 | XP_022269359.1 | 2 - 3 | 440 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
