Assay Details
| Gene Symbol: MSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629930.3 | XP_005629987.1 | 6 - 7 | 1167 | 125 | |
| XM_843168.4 | XP_848261.1 | 6 - 7 | 1169 | 125 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MSR1
Genomic Map