Assay Details
Gene Symbol: KASH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616313.2 | XP_005616370.1 | 5 - 6 | 675 | 74 | |
XM_014118902.2 | XP_013974377.1 | 5 - 6 | 675 | 74 | ||
XM_014118904.2 | XP_013974379.1 | 5 - 6 | 675 | 74 | ||
XM_014118907.2 | XP_013974382.1 | 5 - 6 | 675 | 74 | ||
XM_014118909.2 | XP_013974384.1 | 5 - 6 | 675 | 74 | ||
XM_022423878.1 | XP_022279586.1 | 5 - 6 | 675 | 74 | ||
XM_022423885.1 | XP_022279593.1 | 5 - 6 | 512 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
