Assay Details
| Gene Symbol: ARHGAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641813.3 | XP_005641870.1 | 4 - 5 | 783 | 134 | |
| XM_005641814.3 | XP_005641871.1 | 5 - 6 | 794 | 134 | ||
| XM_014111355.2 | XP_013966830.1 | 5 - 6 | 789 | 134 | ||
| XM_022416069.1 | XP_022271777.1 | 5 - 6 | 776 | 134 | ||
More Information
Set Membership: |
|

ARHGAP36
Genomic Map