Assay Details
| Gene Symbol: C7H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434909.4 | XP_003434957.1 | 3 - 4 | 1089 | 167 | |
| XM_005622845.3 | XP_005622902.1 | 4 - 5 | 1318 | 167 | ||
| XM_005622846.3 | XP_005622903.1 | 4 - 5 | 1339 | 167 | ||
| XM_847758.5 | XP_852851.1 | 4 - 5 | 1219 | 167 | ||
More Information
Set Membership: |
|

C7H18orf25
Genomic Map