Assay Details
| Gene Symbol: C9H17orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624996.3 | XP_005625053.1 | 1 - 2 | 162 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9H17orf50| Gene Symbol: C9H17orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624996.3 | XP_005625053.1 | 1 - 2 | 162 | 106 | |
Set Membership: |
|