Assay Details
| Gene Symbol: IGSF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639517.3 | XP_005639574.1 | 6 - 7 | 920 | 117 | |
| XM_005639518.2 | XP_005639575.1 | 6 - 7 | 869 | 117 | ||
| XM_005639520.3 | XP_005639577.1 | 6 - 7 | 935 | 117 | ||
| XM_005639521.2 | XP_005639578.1 | 6 - 7 | 1279 | 117 | ||
| XM_022414040.1 | XP_022269748.1 | 6 - 7 | 886 | 117 | ||
| XM_022414041.1 | XP_022269749.1 | 6 - 7 | 861 | 117 | ||
| XM_022414042.1 | XP_022269750.1 | 5 - 6 | 800 | 117 | ||
| XM_022414043.1 | XP_022269751.1 | 5 - 6 | 719 | 117 | ||
| XM_022414044.1 | XP_022269752.1 | 6 - 7 | 945 | 117 | ||
| XM_022414045.1 | XP_022269753.1 | 6 - 7 | 829 | 117 | ||
More Information
Set Membership: |
|

IGSF11
Genomic Map