Assay Details
| Gene Symbol: MARCHF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422807.1 | XP_022278515.1 | 2414 | 83 | ||
| XM_022422808.1 | XP_022278516.1 | 2346 | 83 | |||
| XM_022422809.1 | XP_022278517.1 | 2394 | 83 | |||
| XM_022422810.1 | XP_022278518.1 | 2411 | 83 | |||
| XM_022422811.1 | XP_022278519.1 | 2413 | 83 | |||
| XM_022422812.1 | XP_022278520.1 | 2413 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MARCHF10
Genomic Map