Assay Details
| Gene Symbol: SLC23A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109957.1 | NP_001103427.1 | 984 | 113 | ||
| XM_005617236.2 | XP_005617293.2 | 1134 | 113 | |||
| GenBank mRNA | EU125351.1 | - | 984 | 113 | ||
More Information
Set Membership: |
|

SLC23A1
Genomic Map