Assay Details
Gene Symbol: KCNJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619563.3 | XP_005619620.1 | 964 | 128 | ||
XM_014113290.2 | XP_013968765.1 | 729 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
