Assay Details
| Gene Symbol: KCNJ15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638910.3 | XP_005638967.1 | 4 | 1678 | 153 | |
| XM_014109822.2 | XP_013965297.1 | 5 | 1101 | 153 | ||
| XM_014109823.2 | XP_013965298.1 | 4 | 1685 | 153 | ||
| XM_014109825.2 | XP_013965300.1 | 5 | 1094 | 153 | ||
| XM_014109827.2 | XP_013965302.1 | 3 | 1630 | 153 | ||
| XM_014109828.2 | XP_013965303.1 | 4 | 1104 | 153 | ||
| XM_014109829.2 | XP_013965304.1 | 4 | 1048 | 153 | ||
| XM_014109830.2 | XP_013965305.1 | 4 | 996 | 153 | ||
| XM_014109831.2 | XP_013965306.1 | 4 | 1192 | 153 | ||
| XM_022413188.1 | XP_022268896.1 | 5 | 1175 | 153 | ||
| XM_022413189.1 | XP_022268897.1 | 5 | 1124 | 153 | ||
| XM_022413190.1 | XP_022268898.1 | 4 | 1041 | 153 | ||
| XM_022413191.1 | XP_022268899.1 | 4 | 980 | 153 | ||
| XM_022413192.1 | XP_022268900.1 | 4 | 867 | 153 | ||
| XM_022413193.1 | XP_022268901.1 | 3 | 999 | 153 | ||
| XM_022413194.1 | XP_022268902.1 | 3 | 1142 | 153 | ||
| XM_544884.6 | XP_544884.2 | 3 | 948 | 153 | ||
More Information
Set Membership: |
|

KCNJ15
Genomic Map