Assay Details
| Gene Symbol: GTF2A1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204843.1 | NP_001191772.1 | 2 - 3 | 136 | 128 | |
| XM_022423974.1 | XP_022279682.1 | 2 - 3 | 298 | 128 | ||
| GenBank mRNA | BM537539.1 | - | 2 - 3 | 140 | 128 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GTF2A1L
Genomic Map