Assay Details
Gene Symbol: REM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623216.3 | XP_005623273.1 | 5 - 6 | 4896 | 168 | |
XM_005623217.3 | XP_005623274.1 | 4 - 5 | 4848 | 168 | ||
XM_014115750.2 | XP_013971225.1 | 5 - 6 | 4893 | 168 | ||
XM_547725.5 | XP_547725.2 | 4 - 5 | 4845 | 168 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
