Assay Details
| Gene Symbol: SLC35B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127499.1 | NP_001120971.1 | 2 - 3 | 360 | 157 | |
| GenBank mRNA | EF568109.1 | - | 2 - 3 | 360 | 157 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC35B2
Genomic Map