Assay Details
Gene Symbol: RNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634528.3 | XP_005634585.1 | 2 - 3 | 277 | 94 | |
XM_005634530.3 | XP_005634587.1 | 2 - 3 | 252 | 94 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
