Assay Details
Gene Symbol: CYP26C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637866.1 | XP_005637923.1 | 2 - 3 | 426 | 174 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
