Assay Details
| Gene Symbol: E2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630127.2 | XP_005630184.1 | 516 | 153 | ||
| XM_022404453.1 | XP_022260161.1 | 372 | 153 | |||
| XM_022404454.1 | XP_022260162.1 | 438 | 153 | |||
| XM_847370.4 | XP_852463.1 | 645 | 153 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022404454.1 transcripts.
Set Membership: |
|

E2F6
Genomic Map