Assay Details
| Gene Symbol: LOC611401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635169.3 | XP_005635226.1 | 4 - 5 | 480 | 93 | |
| XM_849066.6 | XP_854159.3 | 4 - 5 | 568 | 93 | ||
| GenBank mRNA | CX989699.1 | - | 2 - 3 | 496 | 93 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC611401
Genomic Map