Assay Details
| Gene Symbol: IER5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625249.3 | XP_005625306.1 | 1 | 252 | 82 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  3' Most  Detect Genomic DNA  Recommended Assay  Canis familiaris  Both primers and probe map within a single exon | 
 Back To Top
 Back To Top
 IER5L
IER5L Genomic Map
Genomic Map Back To Top
 Back To Top