Assay Details
| Gene Symbol: SLC25A44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421263.1 | XP_022276971.1 | 2 | 666 | 114 | |
| XM_022421264.1 | XP_022276972.1 | 2 | 668 | 114 | ||
| XM_022421265.1 | XP_022276973.1 | 2 | 672 | 114 | ||
| XM_547537.6 | XP_547537.3 | 2 | 670 | 114 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC25A44
Genomic Map