Assay Details
| Gene Symbol: KAT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114363.2 | XP_013969838.1 | 406 | 72 | ||
| XM_536904.5 | XP_536904.2 | 521 | 72 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KAT8
Genomic Map