Assay Details
| Gene Symbol: FUT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012647.1 | NP_001012665.1 | 362 | 98 | ||
| XM_005629756.3 | XP_005629813.1 | 663 | 98 | |||
| XM_005629757.3 | XP_005629814.1 | 753 | 98 | |||
| GenBank mRNA | AJ879585.1 | - | 362 | 98 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FUT10
Genomic Map