Assay Details
| Gene Symbol: TSC22D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639801.1 | XP_003639849.1 | 2 - 3 | 468 | 78 | |
| XM_005633911.3 | XP_005633968.1 | 2 - 3 | 163 | 78 | ||
| XM_022408317.1 | XP_022264025.1 | 3 - 4 | 3367 | 78 | ||
| XM_022408320.1 | XP_022264028.1 | 3 - 4 | 229 | 78 | ||
| XM_534128.6 | XP_534128.3 | 2 - 3 | 3197 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005633911.3, XM_022408320.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TSC22D1
Genomic Map