Assay Details
| Gene Symbol: KCTD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419878.1 | XP_022275586.1 | 1 | 358 | 126 | |
| XM_547062.6 | XP_547062.1 | 1 | 358 | 126 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KCTD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419878.1 | XP_022275586.1 | 1 | 358 | 126 | |
| XM_547062.6 | XP_547062.1 | 1 | 358 | 126 | ||
Set Membership: |
|