Assay Details
| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620622.3 | XP_005620679.1 | 1 | 291 | 150 | |
| XM_536760.6 | XP_536760.2 | 1 | 291 | 150 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EMC8
Genomic Map