Assay Details
Gene Symbol: HTR1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627599.3 | XP_005627656.1 | 2 | 1111 | 134 | |
XM_005627601.3 | XP_005627658.1 | 3 | 1163 | 134 | ||
XM_022426325.1 | XP_022282033.1 | 3 | 680 | 134 | ||
XM_022426326.1 | XP_022282034.1 | 3 | 783 | 134 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
