Assay Details
| Gene Symbol: CCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005261.1 | NP_001005261.1 | 1 | 1012 | 83 | |
| XM_005632317.3 | XP_005632374.1 | 2 | 1287 | 83 | ||
| GenBank mRNA | AB183193.1 | - | 1 | 1012 | 83 | |
| AY675237.1 | - | 1 | 1012 | 83 | ||
More Information
Set Membership: |
|

CCR3
Genomic Map